This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01197719_g1
See other SUMF2 GE Assays ›
Gene Symbol:
Sumf2
Gene Name:
sulfatase modifying factor 2
Gene Aliases:
2610040F05Rik
Chromosome Location:
Chr.5: 129846990 - 129863951 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01197719_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sumf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026445.2 NP_080721.1 4 - 5 411 77
GenBank mRNA AK013017.1 - 4 - 5 391 77
AK076022.1 - 4 - 5 417 77
AK138300.1 - 4 - 5 402 77
AK157784.1 - 4 - 5 412 77
AK166216.1 - 4 - 5 390 77
BC104404.1 - 2 - 3 520 77
BC104405.1 - 2 - 3 520 77
BC119479.1 - 2 - 3 520 77
BC119480.2 - 2 - 3 520 40050522 77

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sumf2

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein binding
protein homodimerization activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500013695.mm.1 TC0500001514.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500013695.mm.1 NM_026445.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500013695.mm.1 NM_026445.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-tks2p:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0