This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01197720_m1
See other SUMF2 GE Assays ›
Gene Symbol:
Sumf2
Gene Name:
sulfatase modifying factor 2
Gene Aliases:
2610040F05Rik
Chromosome Location:
Chr.5: 129846990 - 129863951 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01197720_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sumf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026445.2 NP_080721.1 5 - 6 558 93
GenBank mRNA AK013017.1 - 5 - 6 538 93
AK076022.1 - 5 - 6 564 93
AK138300.1 - 5 - 6 549 93
AK140169.1 - 6 - 7 573 93
AK157784.1 - 5 - 6 559 93
BC104404.1 - 3 - 4 667 93
BC104405.1 - 3 - 4 667 93
BC119479.1 - 3 - 4 667 93
BC119480.2 - 3 - 4 667 40050522 93

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sumf2

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein binding
protein homodimerization activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0