This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01197762_g1
See other CCT6A GE Assays ›
Gene Symbol:
Cct6a
Gene Name:
chaperonin containing Tcp1, subunit 6a (zeta)
Gene Aliases:
CCT-zeta, CCT-zeta-1, Cct6, Cctz-1, TCP-1-zeta
Chromosome Location:
Chr.5: 129787356 - 129794500 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01197762_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cct6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009838.2 NP_033968.1 694 102
GenBank mRNA AK077916.1 - 647 102
AK134428.1 - 647 102
AK145522.1 - 653 102
AK147130.1 - 691 102
AK149757.1 - 1053 102
AK150977.1 - 648 102
AK159354.1 - 648 102
AK159785.1 - 668 102
AK159975.1 - 694 102
AK160067.1 - 648 102
AK164312.1 - 647 102
AK165612.1 - 650 102
AK166749.1 - 647 102
AK166795.1 - 677 102
AK166870.1 - 645 102
AK166887.1 - 646 102
AK166929.1 - 646 102
AK167007.1 - 676 102
AK167076.1 - 645 102
AK167082.1 - 643 102
AK167164.1 - 653 102
AK167180.1 - 680 102
AK167307.1 - 648 102
AK167669.1 - 677 102
AK167851.1 - 648 102
AK167994.1 - 643 102
AK168065.1 - 643 102
AK168940.1 - 677 102
AK169033.1 - 627 102
AK204878.1 - 143 102
BC018459.1 - 258 3597455 102
BC055043.1 - 656 102
BC069917.1 - 649 6394714 102
BC094352.1 - 620 6809215 102
Z31557.1 - 665 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001512.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cct6a

Panther Classification:

Molecular Function -

chaperonin

Gene Ontology Categories:

Function(s) Process(es)

protein folding
binding of sperm to zona pellucida
toxin transport
nucleotide binding
protein binding
ATP binding
poly(A) RNA binding
unfolded protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500013669.mm.1 NM_009838.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500013669.mm.1 TC0500001512.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline