This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01197937_m1
See other RBPMS GE Assays ›
Gene Symbol:
Rbpms
Gene Name:
RNA binding protein gene with multiple splicing
Gene Aliases:
2010300K22Rik, 2700019M19Rik, AU017537, RBP-MS, hermes
Chromosome Location:
Chr.8: 33782644 - 33929895 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01197937_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbpms
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042674.2 NP_001036139.1 5 - 6 718 70
NM_001042675.2 NP_001036140.1 5 - 6 718 70
NM_019733.3 NP_062707.1 5 - 6 718 70
GenBank mRNA AF148511.1 - 5 - 6 667 70
AK012265.1 - 5 - 6 661 70
AK012586.1 - 5 - 6 661 70
AK041118.1 - 5 - 6 662 70
BC030397.1 - 5 - 6 679 4225078 70
BY708385.1 - 2 - 3 212 70
CK030989.1 - 5 - 6 694 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002012.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbpms

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of pathway-restricted SMAD protein phosphorylation
positive regulation of SMAD protein import into nucleus
regulation of nucleic acid-templated transcription
nucleotide binding
nucleic acid binding
transcription coactivator activity
RNA binding
poly(A) binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800015787.mm.1 NM_001042674.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800015787.mm.1 NM_001042675.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800015787.mm.1 NM_019733.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800015787.mm.1 TC0800002012.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800015788.mm.1 NM_001042674.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800015788.mm.1 NM_001042675.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800015788.mm.1 NM_019733.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800015788.mm.1 TC0800002012.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline