This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01197978_m1
See other GTF2E2 GE Assays ›
Gene Symbol:
Gtf2e2
Gene Name:
general transcription factor II E, polypeptide 2 (beta subunit)
Gene Aliases:
34kDa, AI462509, C330006J08Rik
Chromosome Location:
Chr.8: 33731914 - 33777173 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01197978_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf2e2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026584.3 NP_080860.3 1 - 2 58 73
GenBank mRNA AK088342.1 - 1 - 2 83 73
AK146803.1 - 1 - 2 45 73
BC020016.1 - 1 - 2 38 3501113 73
BY256665.1 - 1 - 2 59 73

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK088342.1, AK146803.1, BC020016.1, NM_026584.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gtf2e2

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
DNA binding
protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800002960.mm.1 NM_026584.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800002960.mm.1 NM_026584.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800002960.mm.1 TC0800000396.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 452189 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline