This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01198306_m1
See other 4930467D21RIK GE Assays ›
Gene Symbol:
4930467D21Rik
Gene Name:
RIKEN cDNA 4930467D21 gene
Gene Aliases:
4930448L09Rik, BB013444
Chromosome Location:
Chr.5: 97392417 - 97588126 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01198306_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930467D21Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045981.1 4 - 5 332 70
GenBank mRNA AK015518.1 - 4 - 5 309 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000940.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500008123.mm.1 NR_045981.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500008123.mm.1 NR_045981.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500008123.mm.1 TC0500000940.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-crt6c:80/100.66.78.36:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline