Assay Details
| Gene Symbol: Rpl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011290.5 | NP_035420.2 | 3 - 4 | 410 | 57 | |
| GenBank mRNA | AK002248.1 | - | 3 - 4 | 375 | 57 | |
| AK002648.1 | - | 3 - 4 | 372 | 57 | ||
| AK010918.1 | - | 2 - 3 | 370 | 57 | ||
| AK088472.1 | - | 2 - 3 | 376 | 57 | ||
| AK135175.1 | - | 3 - 4 | 379 | 57 | ||
| AK148479.1 | - | 3 - 4 | 378 | 57 | ||
| AK150540.1 | - | 3 - 4 | 382 | 57 | ||
| AK151022.1 | - | 3 - 4 | 382 | 57 | ||
| AK151344.1 | - | 3 - 4 | 382 | 57 | ||
| AK151759.1 | - | 3 - 4 | 382 | 57 | ||
| AK153208.1 | - | 3 - 4 | 411 | 57 | ||
| AK153507.1 | - | 3 - 4 | 382 | 57 | ||
| AK167439.1 | - | 3 - 4 | 382 | 57 | ||
| BC062880.1 | - | 2 - 3 | 364 | 5692572 | 57 | |
| BC106121.1 | - | 2 - 3 | 363 | 3979296 | 57 | |
| X81987.1 | - | 2 - 3 | 358 | 57 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24043184 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24043182 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24043181 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24043179 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043639 | AK153507.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043639 | CJ316043.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043639 | AK150540.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043639 | AK151022.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043639 | AK151344.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990859 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052634 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042426 | AK153507.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042426 | AK167439.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042426 | BC062880.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042426 | BC106121.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042426 | X81987.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 382311 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 583572 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 859652 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146300 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043639 | AK153507.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043639 | CJ316043.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043639 | AK150540.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043639 | AK151022.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043639 | AK151344.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052634 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990859 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042426 | AK153507.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042426 | AK167439.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042426 | BC062880.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042426 | BC106121.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042426 | X81987.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P104918 | AK153507.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P104918 | AK167439.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P104918 | AK198216.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P104918 | BC062880.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P104918 | BC106121.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P23041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24043170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24043199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24043175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24043179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24043197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24043181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24043193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24043176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24043195 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24043192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24043205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24043190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24043191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24043173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24043200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24043194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24043183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24043186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24043185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24043202 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146300 | - | - |
| Whole Mouse Genome G4122A | A_52_P23041 | - | - |
| Whole Mouse Genome G4122A | A_52_P104918 | AK153507.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P104918 | AK167439.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P104918 | AK198216.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P104918 | BC062880.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P104918 | BC106121.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146300 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04607977 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04607975 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04607966 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04607968 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04607970 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04212522 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24043182 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24043184 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24043181 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24043179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24043248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043207 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04593009 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043225 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04760569 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04356257 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04607980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04172564 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043259 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04893922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04296661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043229 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04767828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04709799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043205 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04893924 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043191 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043193 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04607974 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043199 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043168 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04220121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24043170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04476747 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10356352 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525189 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525188 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525192 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525193 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525190 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366302 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525191 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525194 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416546_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10356351 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10366301 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10525187 | - | - |
| Mouse Genome 430 2.0 Array | 1416546_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1416546_a_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500011823.mm.1 | NM_011290.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011823.mm.1 | NM_011290.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011823.mm.1 | TC0500001353.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500011810.mm.1 | NM_011290.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011810.mm.1 | NM_011290.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011810.mm.1 | TC0500001353.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500011814.mm.1 | NM_011290.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011814.mm.1 | NM_011290.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011814.mm.1 | TC0500001353.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500011816.mm.1 | NM_011290.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011816.mm.1 | NM_011290.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011816.mm.1 | TC0500001353.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500011819.mm.1 | NM_011290.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011819.mm.1 | NM_011290.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011819.mm.1 | TC0500001353.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500011821.mm.1 | NM_011290.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011821.mm.1 | NM_011290.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011821.mm.1 | TC0500001353.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416546_PM_a_at | - | - |
| Mouse Expression Set 430 | 1416546_a_at | - | - |
| Murine Genome U74v2 Set | 96962_at | - | - |
| Murine Genome U74v2 Set | 96962_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416546_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416546_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416546_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416546_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 899718 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6040735 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006040735 | - | - |
| mouse-6 v1.1 (Illumina) | scl019988.6_33-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6040735 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl019988.6_33-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6040735 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2855066 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2855066 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl019988.6_33-S | - | - |

Rpl6
Genomic Map