This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01198502_m1
See other PTPN11 GE Assays ›
Gene Symbol:
Ptpn11
Gene Name:
protein tyrosine phosphatase, non-receptor type 11
Gene Aliases:
2700084A17Rik, AW536184, PTP1D, PTP2C, SAP-2, SH-PTP2, SH-PTP3, SHP-2, Shp2, Syp
Chromosome Location:
Chr.5: 121130533 - 121191397 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01198502_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptpn11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109992.1 NP_001103462.1 4 - 5 705 97
NM_011202.3 NP_035332.1 4 - 5 705 97
GenBank mRNA AK011468.1 - 4 - 5 667 97
AK147354.1 - 4 - 5 676 97
AK159501.1 - 4 - 5 705 97
AK159587.1 - 4 - 5 705 97
AK163809.1 - 1 - 2 133 97
BC028513.1 - 4 - 5 655 97
BC057398.1 - 4 - 5 637 6409640 97
BC059278.1 - 4 - 5 683 2866038 97
D84372.1 - 4 - 5 581 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003184.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptpn11

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

DNA damage checkpoint
activation of MAPK activity
protein dephosphorylation
lipid metabolic process
triglyceride metabolic process
epidermal growth factor receptor signaling pathway
integrin-mediated signaling pathway
axonogenesis
brain development
heart development
hormone-mediated signaling pathway
positive regulation of signal transduction
dephosphorylation
platelet formation
microvillus organization
abortive mitotic cell cycle
regulation of cell adhesion mediated by integrin
negative regulation of cell adhesion mediated by integrin
multicellular organism growth
organ growth
peptidyl-tyrosine dephosphorylation
megakaryocyte development
atrioventricular canal development
ERBB signaling pathway
regulation of multicellular organism growth
hormone metabolic process
glucose homeostasis
positive regulation of mitotic cell cycle
negative regulation of insulin secretion
regulation of protein export from nucleus
positive regulation of hormone secretion
negative regulation of hormone secretion
platelet-derived growth factor receptor signaling pathway
neurotrophin TRK receptor signaling pathway
ephrin receptor signaling pathway
multicellular organismal reproductive process
genitalia development
inner ear development
homeostasis of number of cells within a tissue
negative regulation of cortisol secretion
negative regulation of growth hormone secretion
face morphogenesis
intestinal epithelial cell migration
positive regulation of ERK1 and ERK2 cascade
positive regulation of glucose import in response to insulin stimulus
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
non-membrane spanning protein tyrosine phosphatase activity
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
hydrolase activity
phosphatase activity
protein domain specific binding
receptor tyrosine kinase binding
D1 dopamine receptor binding
phospholipase binding
insulin receptor substrate binding
peptide hormone receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500028331.mm.1 TC0500003184.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500028331.mm.1 NM_001109992.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500028331.mm.1 NM_001109992.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500028331.mm.1 NM_011202.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500028331.mm.1 NM_011202.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0