This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01198658_m1
See other ARHGAP19 GE Assays ›
Gene Symbol:
Arhgap19
Gene Name:
Rho GTPase activating protein 19
Gene Aliases:
4933411B03Rik, AI120128, AW546831, mFLJ00368
Chromosome Location:
Chr.19: 41766588 - 41802084 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01198658_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163495.1 NP_001156967.1 4 - 5 670 97
NM_027667.3 NP_081943.2 4 - 5 670 97
GenBank mRNA AK016763.1 - 4 - 5 636 97
AK044841.1 - 4 - 5 621 97
AK164477.1 - 4 - 5 632 97
AK220317.1 - 4 - 5 606 97
BC086680.1 - 4 - 5 618 30670200 97
BC139195.1 - 4 - 5 670 8862217 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001510.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgap19

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
molecular_function
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900013264.mm.1 NM_001163495.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013264.mm.1 NM_001163495.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013264.mm.1 NM_027667.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013264.mm.1 NM_027667.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900013264.mm.1 TC1900001510.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900013265.mm.1 NM_001163495.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013265.mm.1 NM_001163495.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013265.mm.1 NM_027667.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013265.mm.1 NM_027667.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900013265.mm.1 TC1900001510.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0