This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01198659_g1
See other ARHGAP19 GE Assays ›
Gene Symbol:
Arhgap19
Gene Name:
Rho GTPase activating protein 19
Gene Aliases:
4933411B03Rik, AI120128, AW546831, mFLJ00368
Chromosome Location:
Chr.19: 41766588 - 41802084 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01198659_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163495.1 NP_001156967.1 5 - 6 898 152
NM_027667.3 NP_081943.2 5 - 6 898 152
GenBank mRNA AK016763.1 - 5 - 6 864 152
AK044841.1 - 5 - 6 849 152
AK164477.1 - 5 - 6 860 152
AK220317.1 - 5 - 6 834 152
BC086680.1 - 5 - 6 846 30670200 152
BC139195.1 - 5 - 6 898 8862217 152

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgap19

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
molecular_function
GTPase activator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0