This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01198662_m1
See other ARHGAP19 GE Assays ›
Gene Symbol:
Arhgap19
Gene Name:
Rho GTPase activating protein 19
Gene Aliases:
4933411B03Rik, AI120128, AW546831, mFLJ00368
Chromosome Location:
Chr.19: 41766588 - 41802084 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01198662_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163495.1 NP_001156967.1 8 - 9 1239 66
NM_027667.3 NP_081943.2 8 - 9 1239 66
GenBank mRNA AK016763.1 - 8 - 9 1160 66
AK044841.1 - 8 - 9 1190 66
AK164477.1 - 8 - 9 1201 66
AK220317.1 - 8 - 9 1175 66
BC086680.1 - 8 - 9 1187 30670200 66
BC139195.1 - 8 - 9 1239 8862217 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001510.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgap19

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
molecular_function
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900013259.mm.1 NM_001163495.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013259.mm.1 NM_001163495.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013259.mm.1 NM_027667.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013259.mm.1 NM_027667.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900013259.mm.1 TC1900001510.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900013260.mm.1 NM_001163495.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013260.mm.1 NM_001163495.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013260.mm.1 NM_027667.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013260.mm.1 NM_027667.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900013260.mm.1 TC1900001510.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900013261.mm.1 NM_001163495.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013261.mm.1 NM_001163495.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013261.mm.1 NM_027667.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013261.mm.1 NM_027667.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900013261.mm.1 TC1900001510.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900013263.mm.1 NM_001163495.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013263.mm.1 NM_001163495.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013263.mm.1 NM_027667.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013263.mm.1 NM_027667.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900013263.mm.1 TC1900001510.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0