This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01198718_m1
See other SLC7A2 GE Assays ›
Gene Symbol:
Slc7a2
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 2
Gene Aliases:
20.5, AI158848, Atrc2, CAT-2, Cat2, Tea
Chromosome Location:
Chr.8: 40862367 - 40922070 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01198718_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007514.3 NP_031540.2 7 - 8 1397 71
GenBank mRNA BC021653.1 - 7 - 8 1392 71
BC127082.1 - 7 - 8 1313 40111372 71
DQ086834.1 - 7 - 8 1352 71
L03290.1 - 6 - 7 1199 71
L11600.1 - 6 - 7 1199 71
L29006.1 - 6 - 7 1722 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800000482.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc7a2

Panther Classification:

Molecular Function -

amino acid transporter

Gene Ontology Categories:

Function(s) Process(es)

nitric oxide production involved in inflammatory response
amino acid transmembrane transport
nitric oxide biosynthetic process
transport
amino acid transport
arginine transport
macrophage activation
regulation of macrophage activation
regulation of inflammatory response
transmembrane transport
L-arginine import across plasma membrane
L-lysine import across plasma membrane
L-ornithine import across plasma membrane
L-arginine transport
L-ornithine transmembrane transport
L-arginine transmembrane transport
arginine transmembrane transport
L-ornithine transmembrane transporter activity
high-affinity arginine transmembrane transporter activity
high-affinity lysine transmembrane transporter activity
amino acid transmembrane transporter activity
arginine transmembrane transporter activity
L-lysine transmembrane transporter activity
antiporter activity
L-arginine transmembrane transporter activity
low-affinity L-arginine transmembrane transporter activity
high-affinity L-ornithine transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline