This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01198719_m1
See other SLC7A2 GE Assays ›
Gene Symbol:
Slc7a2
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 2
Gene Aliases:
20.5, AI158848, Atrc2, CAT-2, Cat2, Tea
Chromosome Location:
Chr.8: 40862367 - 40922070 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01198719_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK155558.1 - 8 - 9 1478 61
AK171369.1 - 8 - 9 1474 61
BC021653.1 - 8 - 9 1494 61
BC127082.1 - 8 - 9 1415 40111372 61
DQ086834.1 - 8 - 9 1454 61
L11600.1 - 7 - 8 1301 61
L29006.1 - 7 - 8 1824 61
M62838.1 - 8 - 9 1415 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800000482.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

amino acid transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline