This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01198878_g1
See other TMEM175 GE Assays ›
Gene Symbol:
Tmem175
Gene Name:
transmembrane protein 175
Gene Aliases:
3010001K23Rik, AI504381
Chromosome Location:
Chr.5: 108629801 - 108647770 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01198878_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem175
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163531.1 NP_001157003.1 6 - 7 563 102
NM_001163532.1 NP_001157004.1 6 - 7 566 102
NM_028223.3 NP_082499.3 6 - 7 673 102
GenBank mRNA AI526594.1 - 6 - 7 537 102
AK013879.1 - 6 - 7 496 102
AK034579.1 - 6 - 7 542 102
AK038865.1 - 6 - 7 533 102
AK049952.1 - 6 - 7 607 102
AK082765.1 - 6 - 7 529 102
AK170273.1 - 6 - 7 516 102
BC005542.1 - 6 - 7 481 3590497 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001110.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmem175

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500009466.mm.1 TC0500001110.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500009466.mm.1 NM_001163531.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009466.mm.1 NM_001163531.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009466.mm.1 NM_001163532.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009466.mm.1 NM_001163532.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009466.mm.1 NM_028223.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009466.mm.1 NM_028223.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0