This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01199461_m1
See other ABCC2 GE Assays ›
Gene Symbol:
Abcc2
Gene Name:
ATP-binding cassette, sub-family C (CFTR/MRP), member 2
Gene Aliases:
AI173996, Abc30, Cmoat, Mrp2, cMRP
Chromosome Location:
Chr.19: 43782308 - 43838332 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01199461_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Abcc2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013806.2 NP_038834.2 23 - 24 3347 58
GenBank mRNA AF282772.1 - 23 - 24 3251 58
AF282773.1 - 23 - 24 3251 58

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Abcc2

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

drug transmembrane transport
prostaglandin transport
cellular chloride ion homeostasis
response to drug
response to estrogen
thyroid hormone transport
protein binding
protein domain specific binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0