This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01199503_m1
See other P2RX7 GE Assays ›
Gene Symbol:
P2rx7
Gene Name:
purinergic receptor P2X, ligand-gated ion channel, 7
Gene Aliases:
AI467586, P2X(7), P2X7R
Chromosome Location:
Chr.5: 122643911 - 122691432 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01199503_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P2rx7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001038839.2 NP_001033928.1 7 - 8 903 69
NM_001038845.2 NP_001033934.1 7 - 8 903 69
NM_001284402.1 NP_001271331.1 7 - 8 731 69
NM_011027.3 NP_035157.2 7 - 8 903 69
GenBank mRNA AJ009823.1 - 7 - 8 751 69
AK144585.1 - 7 - 8 904 69
AK145050.1 - 7 - 8 758 69
AK145211.1 - 7 - 8 767 69
AK171132.1 - 7 - 8 875 69
BC128303.1 - 7 - 8 747 69
BC128304.1 - 7 - 8 747 69
BC141120.1 - 7 - 8 770 9055732 69
FJ436444.1 - 7 - 8 731 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001399.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene P2rx7

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
cell morphogenesis
phagolysosome assembly
positive regulation of T cell mediated cytotoxicity
positive regulation of protein phosphorylation
protein phosphorylation
membrane protein ectodomain proteolysis
phospholipid transfer to membrane
transport
ion transport
cation transport
calcium ion transport
cell volume homeostasis
membrane budding
inflammatory response
mitochondrion organization
plasma membrane organization
response to mechanical stimulus
response to bacterium
response to organic substance
response to zinc ion
gene expression
positive regulation of calcium ion transport into cytosol
positive regulation of gene expression
programmed cell death
positive regulation of glutamate secretion
positive regulation of gamma-aminobutyric acid secretion
response to organic cyclic compound
synaptic vesicle exocytosis
protein processing
phospholipid scrambling
sensory perception of pain
cytolysis
positive regulation of bone mineralization
cellular response to extracellular stimulus
bleb assembly
positive regulation of prostaglandin secretion
response to lipopolysaccharide
positive regulation of interleukin-1 beta production
positive regulation of interleukin-6 production
collagen metabolic process
response to ATP
response to fluid shear stress
positive regulation of ion transmembrane transport
purinergic nucleotide receptor signaling pathway
T cell proliferation
response to drug
T cell homeostasis
positive regulation of apoptotic process
positive regulation of catalytic activity
NAD transport
negative regulation of MAPK cascade
positive regulation of MAPK cascade
multicellular organismal protein catabolic process
phospholipid translocation
positive regulation of ossification
negative regulation of bone resorption
negative regulation of cell volume
positive regulation of glycolytic process
ceramide biosynthetic process
pore complex assembly
skeletal system morphogenesis
homeostasis of number of cells within a tissue
positive regulation of protein secretion
positive regulation of cytokine secretion
positive regulation of interleukin-1 alpha secretion
positive regulation of interleukin-1 beta secretion
defense response to Gram-positive bacterium
release of sequestered calcium ion into cytosol
protein oligomerization
response to calcium ion
response to electrical stimulus
membrane depolarization
positive regulation of mitochondrial depolarization
positive regulation of lymphocyte apoptotic process
cellular response to dsRNA
cellular response to organic cyclic compound
reactive oxygen species metabolic process
extrinsic apoptotic signaling pathway
cation transmembrane transport
positive regulation of bleb assembly
lipopolysaccharide binding
purinergic nucleotide receptor activity
receptor activity
extracellular ATP-gated cation channel activity
ion channel activity
protein binding
ATP binding
channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0