This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01199854_m1
See other PLBD2 GE Assays ›
Gene Symbol:
Plbd2
Gene Name:
phospholipase B domain containing 2
Gene Aliases:
1300012G16Rik, AU019810, P76
Chromosome Location:
Chr.5: 120483893 - 120503623 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01199854_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Plbd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023625.4 NP_076114.2 2 - 3 423 78
GenBank mRNA AF217003.1 - 2 - 3 404 78
AK004973.1 - 2 - 3 426 78
AK029813.1 - 2 - 3 423 78
AK030234.1 - 2 - 3 375 78
AK032930.1 - 2 - 3 419 78
AK042089.1 - 419 78
AK043608.1 - 2 - 3 413 78
AK046937.1 - 2 - 3 413 78
AK150201.1 - 2 - 3 414 78
AK164220.1 - 2 - 3 404 78
AK170387.1 - 2 - 3 415 78
AK170631.1 - 2 - 3 403 78
BC026395.1 - 2 - 3 401 78
BC038605.1 - 2 - 3 404 4921245 78
BC048826.1 - 2 - 3 410 4196604 78
BC060095.1 - 2 - 3 401 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003168.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Plbd2

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
biological_process
lipid catabolic process
molecular_function
phospholipase activity
prenylcysteine methylesterase activity
lipase activity
hydrolase activity
1-oxa-2-oxocycloheptane lactonase activity
sulfolactone hydrolase activity
3,4-dihydrocoumarin hydrolase activity
butyrolactone hydrolase activity
endosulfan lactone lactonase activity
L-ascorbate 6-phosphate lactonase activity
Ser-tRNA(Thr) hydrolase activity
Ala-tRNA(Pro) hydrolase activity
Cys-tRNA(Pro) hydrolase activity
Ser(Gly)-tRNA(Ala) hydrolase activity
all-trans-retinyl-palmitate hydrolase, all-trans-retinol forming activity
retinyl-palmitate esterase activity
mannosyl-oligosaccharide 1,6-alpha-mannosidase activity
mannosyl-oligosaccharide 1,3-alpha-mannosidase activity
methyl indole-3-acetate esterase activity
methyl salicylate esterase activity
methyl jasmonate esterase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500028080.mm.1 NM_023625.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500028080.mm.1 NM_023625.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500028080.mm.1 TC0500003168.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline