This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01200022_g1
See other DRAM2 GE Assays ›
Gene Symbol:
Dram2
Gene Name:
DNA-damage regulated autophagy modulator 2
Gene Aliases:
2010305N14Rik, 2610318G18Rik, AI647667, Tmem77
Chromosome Location:
Chr.3: 106547798 - 106575341 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01200022_g1
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dram2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025582.2 NP_001020753.1 6 - 7 689 90
NM_001286986.1 NP_001273915.1 8 - 9 947 90
NM_001286987.1 NP_001273916.1 7 - 8 825 90
NM_026013.3 NP_080289.1 8 - 9 950 90
GenBank mRNA AK008532.1 - 6 - 7 613 90
AK009940.1 - 8 - 9 887 90
AK012044.1 - 8 - 9 867 90
AK015888.1 - 8 - 9 913 90
AK052824.1 - 7 - 8 759 90
AK172455.1 - 8 - 9 917 90
BC024458.1 - 6 - 7 615 4975302 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001104.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dram2

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
regulation of autophagy
molecular_function

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22402370 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22402369 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22402368 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980746 AK172455.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980746 BC024458.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980746 AK012044.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980746 AK015888.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980746 AK009940.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 411844 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874938 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 788206 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 971843 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290255 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647140 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 564740 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913000 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 662517 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 86088 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 955129 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 944655 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 437588 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 66204 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 508848 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 221243 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 302248 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 81152 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 105316 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125526 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151468 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980746 AK172455.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980746 BC024458.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980746 AK012044.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980746 AK015888.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980746 AK009940.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402384 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402370 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402389 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402399 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402373 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402375 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402381 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402374 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402398 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402397 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402386 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402393 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402382 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402401 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402376 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402379 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402369 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402396 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402391 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402378 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402368 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22402377 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151468 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00720312 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125526 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00720304 - -
Whole Mouse Genome G4122A A_51_P380801 AK172455.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P380801 BC024458.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P380801 AK012044.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P380801 AK052824.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P380801 NM_001025582.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120520 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151468 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125526 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00720304 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02545951 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02545944 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02545940 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22402368 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22402370 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22402369 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402405 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402436 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402457 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402500 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402466 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402421 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402447 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402456 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402368 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402476 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402420 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402398 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402408 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402386 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402498 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402373 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402494 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402430 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402504 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402424 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402517 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402410 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402461 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402513 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402407 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402396 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402375 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402450 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402470 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402502 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402463 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402519 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402417 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402382 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402426 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402493 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402411 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402516 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402454 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402471 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402413 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402512 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402379 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402465 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402508 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402431 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402434 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402490 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402406 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402487 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402443 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402506 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402448 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402459 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402482 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402409 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402384 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402472 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402389 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402391 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402518 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402381 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402505 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402475 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402377 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402412 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402371 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402397 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402415 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402503 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402480 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402477 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402441 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22402403 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402384 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402408 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402434 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402396 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402420 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402371 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402386 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402425 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402401 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402379 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402415 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402409 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402393 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402421 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402373 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402375 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402387 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402398 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402412 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402394 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402391 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402382 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402418 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402410 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02673354 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402441 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402431 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402368 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402413 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402397 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02744481 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402430 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402403 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402389 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402426 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402417 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402411 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402427 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402424 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402407 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402381 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402378 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402388 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402436 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22402377 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10495165 - -
Affymetrix Mouse Gene 1.0 ST Array 10495166 - -
Affymetrix Mouse Gene 1.0 ST Array 10495171 - -
Affymetrix Mouse Gene 1.0 ST Array 10495170 - -
Affymetrix Mouse Gene 1.0 ST Array 10495169 - -
Affymetrix Mouse Gene 1.0 ST Array 10495168 - -
Affymetrix Mouse Gene 1.0 ST Array 10495172 - -
Affymetrix Mouse Gene 1.0 ST Array 10495164 - -
Affymetrix Mouse Gene 1.0 ST Array 10495167 - -
Affymetrix HT MG-430 PM Array Plate 1445545_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1453731_PM_a_at - -
Affymetrix HT MG-430 PM Array Plate 1424782_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10495163 - -
Mouse Genome 430 2.0 Array 1453731_a_at - -
Mouse Genome 430 2.0 Array 1453731_PM_a_at - -
Mouse Genome 430 2.0 Array 1445545_PM_at - -
Mouse Genome 430 2.0 Array 1424782_at - -
Mouse Genome 430 2.0 Array 1424782_PM_at - -
Mouse Genome 430 2.0 Array 1445545_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 67171_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 67171_PM_at - -
Affymetrix Mouse Clariom S Array PSR0300008998.mm.1 NM_001025582.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300008998.mm.1 NM_001286986.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008998.mm.1 NM_001286987.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008998.mm.1 NM_026013.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300008998.mm.1 TC0300001104.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300009000.mm.1 NM_001025582.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300009000.mm.1 NM_001286986.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009000.mm.1 NM_001286987.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009000.mm.1 NM_026013.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300009000.mm.1 TC0300001104.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300009001.mm.1 NM_001025582.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300009001.mm.1 NM_001286986.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009001.mm.1 NM_001286987.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009001.mm.1 NM_026013.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300009001.mm.1 TC0300001104.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300008999.mm.1 NM_001025582.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300008999.mm.1 NM_001286986.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008999.mm.1 NM_001286987.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008999.mm.1 NM_026013.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300008999.mm.1 TC0300001104.mm.1 MTA TC
Mouse Expression Set 430 1453731_a_at - -
Mouse Expression Set 430 1453731_PM_a_at - -
Mouse Expression Set 430 1424782_at - -
Mouse Expression Set 430 1424782_PM_at - -
Mouse Expression Set 430 1445545_PM_at - -
Mouse Expression Set 430 1445545_at - -
Murine Genome U74v2 Set 96688_PM_at - -
Murine Genome U74v2 Set 161540_PM_r_at - -
Murine Genome U74v2 Set 161540_r_at - -
Murine Genome U74v2 Set 96688_at - -
Mouse Genome 430A 2.0 Array 1453731_a_at - -
Mouse Genome 430A 2.0 Array 1453731_PM_a_at - -
Mouse Genome 430A 2.0 Array 1424782_PM_at - -
Mouse Genome 430A 2.0 Array 1424782_at - -
Affymetrix HT Mouse Genome 430A Array 1424782_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1453731_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1424782_at - -
Affymetrix HT Mouse Genome 430A Array 1453731_a_at - -
Affymetrix HT Mouse Genome 430B Array 1445545_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1445545_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 6110324 - -
Illumina mouse-6 v1.0 expression beadchip 100430368 - -
Illumina mouse-6 v1.0 expression beadchip 6110324 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006110324 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1224589 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1224589 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2489135 - -
mouse-6 v1.1 (Illumina) scl22747.11_296-S - -
mouse-6 v1.1 (Illumina) scl0067171.2_285-S - -
Illumina Sentrix mouse-6.v1 bead array 6110324 - -
Illumina Sentrix mouse-6.v1 bead array 100430368 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0067171.2_285-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6110324 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 100430368 - -
MouseRef-8 v2 Expression BeadChip ILMN_3051608 - -
MouseRef-8 v2 Expression BeadChip ILMN_1224589 - -
MouseRef-8 v2 Expression BeadChip ILMN_3127932 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_3051608 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_3127932 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1224589 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0