This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01200171_m1
See other PTCD1 GE Assays ›
Gene Symbol:
Ptcd1
Gene Name:
pentatricopeptide repeat domain 1
Gene Aliases:
1110069M14Rik
Chromosome Location:
Chr.5: 145147378 - 145167119 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01200171_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptcd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133735.2 NP_598496.3 3 - 4 770 102
GenBank mRNA AK088764.1 - 3 - 4 689 102
AK143877.1 - 3 - 4 727 102
AK150514.1 - 3 - 4 727 102
AK151064.1 - 3 - 4 739 101
AK152077.1 - 3 - 4 739 101
AK153366.1 - 3 - 4 739 101
AK171484.1 - 3 - 4 725 102
BC004766.1 - 3 - 4 688 3156304 102
BC019121.1 - 1 - 2 158 102
BC083097.1 - 3 - 4 729 30459747 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003590.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptcd1

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

tRNA processing
biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500032935.mm.1 TC0500003590.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500032935.mm.1 NM_133735.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500032935.mm.1 NM_133735.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0