This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01200385_m1
See other RBPMS GE Assays ›
Gene Symbol:
Rbpms
Gene Name:
RNA binding protein gene with multiple splicing
Gene Aliases:
2010300K22Rik, 2700019M19Rik, AU017537, RBP-MS, hermes
Chromosome Location:
Chr.8: 33782644 - 33929895 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01200385_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbpms
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042674.2 NP_001036139.1 7 - 8 922 108
GenBank mRNA AK041118.1 - 7 - 8 866 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of pathway-restricted SMAD protein phosphorylation
positive regulation of SMAD protein import into nucleus
regulation of nucleic acid-templated transcription
nucleotide binding
nucleic acid binding
transcription coactivator activity
RNA binding
poly(A) binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline