This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01200387_m1
See other RBPMS GE Assays ›
Gene Symbol:
Rbpms
Gene Name:
RNA binding protein gene with multiple splicing
Gene Aliases:
2010300K22Rik, 2700019M19Rik, AU017537, RBP-MS, hermes
Chromosome Location:
Chr.8: 33782644 - 33929895 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01200387_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbpms
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042675.2 NP_001036140.1 6 - 7 856 63
GenBank mRNA AK012265.1 - 6 - 7 799 63
AK012586.1 - 6 - 7 799 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002012.mm.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of pathway-restricted SMAD protein phosphorylation
positive regulation of SMAD protein import into nucleus
regulation of nucleic acid-templated transcription
nucleotide binding
nucleic acid binding
transcription coactivator activity
RNA binding
poly(A) binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800015784.mm.1 NM_001042675.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800015784.mm.1 TC0800002012.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800015785.mm.1 NM_001042674.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800015785.mm.1 NM_001042675.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800015785.mm.1 NM_019733.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800015785.mm.1 TC0800002012.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0