This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01201045_m1
See other PDGFA GE Assays ›
Gene Symbol:
Pdgfa
Gene Name:
platelet derived growth factor, alpha
Gene Aliases:
-
Chromosome Location:
Chr.5: 138976381 - 138999916 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01201045_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pdgfa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008808.3 NP_032834.1 4 - 5 637 92
GenBank mRNA AK017169.1 - 2 - 3 222 92
AY324648.1 - 4 - 5 626 92
BC003817.1 - 4 - 5 637 3495629 92
M29464.1 - 4 - 5 517 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003482.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pdgfa

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
hair follicle development
positive regulation of mesenchymal cell proliferation
multicellular organism development
positive regulation of cell proliferation
response to wounding
organ morphogenesis
negative regulation of phosphatidylinositol biosynthetic process
negative regulation of platelet activation
positive regulation of phosphatidylinositol 3-kinase signaling
regulation of smooth muscle cell migration
cell projection assembly
actin cytoskeleton organization
positive regulation of cell migration
positive regulation of protein autophosphorylation
positive regulation of metanephric mesenchymal cell migration by platelet-derived growth factor receptor-beta signaling pathway
positive regulation of MAP kinase activity
positive regulation of MAPK cascade
skin development
positive regulation of DNA replication
platelet-derived growth factor receptor signaling pathway
positive regulation of fibroblast proliferation
lung alveolus development
digestive tract development
regulation of peptidyl-tyrosine phosphorylation
negative chemotaxis
positive regulation of cell division
positive regulation of protein kinase B signaling
bone development
regulation of branching involved in salivary gland morphogenesis by epithelial-mesenchymal signaling
positive regulation of ERK1 and ERK2 cascade
embryonic lung development
platelet-derived growth factor receptor binding
collagen binding
growth factor activity
protein homodimerization activity
protein heterodimerization activity
platelet-derived growth factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500031842.mm.1 NM_008808.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500031842.mm.1 NM_008808.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500031842.mm.1 TC0500003482.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500031841.mm.1 NM_008808.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500031841.mm.1 NM_008808.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500031841.mm.1 TC0500003482.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline