Assay Details
| Gene Symbol: Ephb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159571.1 | NP_001153043.1 | 3 - 4 | 906 | 64 | |
| NM_010144.6 | NP_034274.4 | 3 - 4 | 906 | 64 | ||
| GenBank mRNA | AK046884.1 | - | 3 - 4 | 844 | 64 | |
| AK049340.1 | - | 3 - 4 | 907 | 64 | ||
| AK132392.1 | - | 3 - 4 | 906 | 64 | ||
| AK143422.1 | - | 875 | 64 | |||
| BC013342.1 | - | 3 - 4 | 566 | 64 | ||
| BC016505.1 | - | 3 - 4 | 858 | 3709926 | 64 | |
| BC090839.1 | - | 3 - 4 | 683 | 64 | ||
| U06834.1 | - | 4 - 5 | 519 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ephb4
Genomic Map