This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01201196_m1
See other GTF2I GE Assays ›
Gene Symbol:
Gtf2i
Gene Name:
general transcription factor II I
Gene Aliases:
6030441I21Rik, BAP-135, Diws1t, GtfII-I, Spin, TFII-I, WBSCR6
Chromosome Location:
Chr.5: 134237832 - 134314760 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01201196_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf2i
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080746.2 NP_001074215.1 1 - 2 376 98
NM_001080747.2 NP_001074216.1 1 - 2 376 98
NM_001080748.2 NP_001074217.1 1 - 2 376 98
NM_001080749.2 NP_001074218.1 1 - 2 376 98
NM_010365.4 NP_034495.2 1 - 2 376 98
GenBank mRNA AK147201.1 - 1 - 2 334 98
BC053044.1 - 1 - 2 345 5704935 98
CD355048.1 - 1 - 2 361 98
CF540006.1 - 1 - 2 343 98

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK147201.1, BC053044.1, NM_001080746.2, NM_001080747.2, NM_001080748.2, NM_001080749.2, NM_010365.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gtf2i

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
embryo development
transition between slow and fast fiber
negative regulation of angiogenesis
negative regulation of cytosolic calcium ion concentration
DNA binding
protein binding
mitogen-activated protein kinase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500030354.mm.1 TC0500003369.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500030354.mm.1 NM_001080746.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030354.mm.1 NM_001080747.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030354.mm.1 NM_001080748.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030354.mm.1 NM_001080749.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030354.mm.1 NM_010365.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500030353.mm.1 TC0500003369.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500030353.mm.1 NM_001080746.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030353.mm.1 NM_001080747.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030353.mm.1 NM_001080748.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030353.mm.1 NM_001080749.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030353.mm.1 NM_010365.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500030343.mm.1 TC0500003369.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500030343.mm.1 NM_001080746.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030343.mm.1 NM_001080747.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030343.mm.1 NM_001080748.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030343.mm.1 NM_001080749.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030343.mm.1 NM_010365.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500030356.mm.1 TC0500003369.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500030356.mm.1 NM_001080746.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030356.mm.1 NM_001080747.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030356.mm.1 NM_001080748.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030356.mm.1 NM_001080749.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030356.mm.1 NM_010365.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500030338.mm.1 TC0500003369.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500030338.mm.1 NM_001080746.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030338.mm.1 NM_001080747.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030338.mm.1 NM_001080748.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030338.mm.1 NM_001080749.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030338.mm.1 NM_010365.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500030341.mm.1 TC0500003369.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500030341.mm.1 NM_001080746.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030341.mm.1 NM_001080747.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030341.mm.1 NM_001080748.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030341.mm.1 NM_001080749.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030341.mm.1 NM_010365.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x8dcn:80/100.66.79.134:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0