This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01201231_m1
See other EIF4H GE Assays ›
Gene Symbol:
Eif4h
Gene Name:
eukaryotic translation initiation factor 4H
Gene Aliases:
AU018978, D5Ertd355e, E430026L18Rik, Ef4h, Wbscr1, Wscr1, mKIAA0038
Chromosome Location:
Chr.5: 134619876 - 134639494 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01201231_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif4h
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK136999.1 - 4 - 5 432 126
AK168872.1 - 4 - 5 464 126
AK169407.1 - 4 - 5 464 126
AK188319.1 - 1 - 2 101 126
BC014796.1 - 4 - 5 459 3962104 126

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0