This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01201278_m1
See other P2RX4 GE Assays ›
Gene Symbol:
P2rx4
Gene Name:
purinergic receptor P2X, ligand-gated ion channel 4
Gene Aliases:
AI504491, AW555605, D5Ertd444e, P2X4
Chromosome Location:
Chr.5: 122707504 - 122729739 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01201278_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P2rx4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011026.2 NP_035156.2 5 - 6 622 68
GenBank mRNA AF089751.1 - 5 - 6 556 68
AJ251459.1 - 5 - 6 536 68
AJ251461.1 - 5 - 6 536 68
AK150351.1 - 5 - 6 612 68
AK162820.1 - 5 - 6 588 68
AK163104.1 - 5 - 6 610 68
BC002099.1 - 5 - 6 69
JX417451.1 - 5 - 6 582 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001401.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene P2rx4

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

regulation of sodium ion transport
nitric oxide biosynthetic process
transport
ion transport
calcium ion transport
signal transduction
regulation of blood pressure
positive regulation of calcium ion transport into cytosol
negative regulation of cardiac muscle hypertrophy
neuronal action potential
sensory perception of pain
response to ATP
ion transmembrane transport
response to fluid shear stress
purinergic nucleotide receptor signaling pathway
vasodilation
positive regulation of calcium-mediated signaling
protein homooligomerization
membrane depolarization
regulation of cardiac muscle contraction
relaxation of cardiac muscle
excitatory postsynaptic potential
cellular response to ATP
apoptotic signaling pathway
cation transmembrane transport
purinergic nucleotide receptor activity
extracellular ATP-gated cation channel activity
receptor binding
ion channel activity
copper ion binding
ATP binding
drug binding
zinc ion binding
cadherin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500012401.mm.1 TC0500001401.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500012401.mm.1 NM_011026.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0