This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01201284_g1
See other PDE6B GE Assays ›
Gene Symbol:
Pde6b
Gene Name:
phosphodiesterase 6B, cGMP, rod receptor, beta polypeptide
Gene Aliases:
Pdeb, r, rd, rd-1, rd1, rd10
Chromosome Location:
Chr.5: 108388373 - 108431743 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01201284_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde6b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008806.2 NP_032832.2 18 - 19 2263 93
GenBank mRNA AK039397.1 - 18 - 19 1975 93
AK044364.1 - 18 - 19 2206 93
BC048247.1 - 18 - 19 2242 93
BC129923.1 - 18 - 19 2193 40111882 93
BC129924.1 - 18 - 19 2192 93
X60133.1 - 18 - 19 2203 93
X87952.1 - 18 - 19 2203 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001103.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pde6b

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
visual perception
detection of light stimulus
GMP metabolic process
response to stimulus
regulation of cytosolic calcium ion concentration
retina development in camera-type eye
3',5'-cyclic-nucleotide phosphodiesterase activity
phosphoric diester hydrolase activity
hydrolase activity
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500009400.mm.1 NM_008806.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009400.mm.1 NM_008806.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500009400.mm.1 TC0500001103.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500009398.mm.1 NM_008806.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009398.mm.1 NM_008806.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500009398.mm.1 TC0500001103.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline