This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01201286_m1
See other PDE6B GE Assays ›
Gene Symbol:
Pde6b
Gene Name:
phosphodiesterase 6B, cGMP, rod receptor, beta polypeptide
Gene Aliases:
Pdeb, r, rd, rd-1, rd1, rd10
Chromosome Location:
Chr.5: 108388373 - 108431743 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01201286_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde6b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008806.2 NP_032832.2 1 - 2 533 70
GenBank mRNA AK039397.1 - 1 - 2 245 70
AK044364.1 - 1 - 2 476 70
BC048247.1 - 1 - 2 512 70
BC058803.1 - 1 - 2 480 70
BC129923.1 - 1 - 2 463 40111882 70
BC129924.1 - 1 - 2 462 70
BU505563.1 - 1 - 2 533 70
X60133.1 - 1 - 2 473 70
X87952.1 - 1 - 2 473 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001103.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pde6b

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
visual perception
detection of light stimulus
GMP metabolic process
response to stimulus
regulation of cytosolic calcium ion concentration
retina development in camera-type eye
3',5'-cyclic-nucleotide phosphodiesterase activity
phosphoric diester hydrolase activity
hydrolase activity
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500009379.mm.1 NM_008806.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009379.mm.1 NM_008806.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500009379.mm.1 TC0500001103.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500009378.mm.1 NM_008806.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009378.mm.1 NM_008806.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500009378.mm.1 TC0500001103.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500009380.mm.1 NM_008806.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009380.mm.1 NM_008806.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500009380.mm.1 TC0500001103.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500009376.mm.1 NM_008806.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009376.mm.1 NM_008806.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500009376.mm.1 TC0500001103.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 912710 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline