This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01201462_m1
See other DAGLB GE Assays ›
Gene Symbol:
Daglb
Gene Name:
diacylglycerol lipase, beta
Gene Aliases:
E330036I19Rik
Chromosome Location:
Chr.5: 143464487 - 143504442 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01201462_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Daglb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144915.3 NP_659164.2 11 - 12 1615 76
GenBank mRNA AK086747.1 - 11 - 12 1527 76
AK087884.1 - 11 - 12 1470 76
AK136372.1 - 11 - 12 1473 76
BC016105.1 - 11 - 12 1511 4921222 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001734.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Daglb

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
neuroblast proliferation
lipid catabolic process
arachidonic acid metabolic process
neurogenesis
neurotransmitter biosynthetic process
phospholipase activity
prenylcysteine methylesterase activity
lipase activity
hydrolase activity
1-oxa-2-oxocycloheptane lactonase activity
sulfolactone hydrolase activity
3,4-dihydrocoumarin hydrolase activity
butyrolactone hydrolase activity
endosulfan lactone lactonase activity
L-ascorbate 6-phosphate lactonase activity
Ser-tRNA(Thr) hydrolase activity
Ala-tRNA(Pro) hydrolase activity
Cys-tRNA(Pro) hydrolase activity
Ser(Gly)-tRNA(Ala) hydrolase activity
metal ion binding
all-trans-retinyl-palmitate hydrolase, all-trans-retinol forming activity
retinyl-palmitate esterase activity
mannosyl-oligosaccharide 1,6-alpha-mannosidase activity
mannosyl-oligosaccharide 1,3-alpha-mannosidase activity
methyl indole-3-acetate esterase activity
methyl salicylate esterase activity
methyl jasmonate esterase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500015939.mm.1 TC0500001734.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500015939.mm.1 NM_144915.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500015939.mm.1 NM_144915.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0