This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01201566_m1
See other ARHGAP24 GE Assays ›
Gene Symbol:
Arhgap24
Gene Name:
Rho GTPase activating protein 24
Gene Aliases:
0610025G21Rik
Chromosome Location:
Chr.5: 102207198 - 102897937 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01201566_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap24
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029270.2 NP_083546.2 2 - 3 664 112
GenBank mRNA AK031879.1 - 2 - 3 251 112
AK052395.1 - 2 - 3 527 112
AK080986.1 - 2 - 3 664 112

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK031879.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgap24

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
signal transduction
multicellular organism development
cell differentiation
negative regulation of Rac protein signal transduction
wound healing, spreading of epidermal cells
activation of GTPase activity
negative regulation of ruffle assembly
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500008420.mm.1 TC0500000997.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500008420.mm.1 NM_029270.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500008420.mm.1 NM_029270.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500008420.mm.1 NM_146161.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500008420.mm.1 NM_146161.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0