This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01201640_m1
See other DTX2 GE Assays ›
Gene Symbol:
Dtx2
Gene Name:
deltex 2 homolog (Drosophila)
Gene Aliases:
2610524D08Rik, AA408415, AU022494, Deltex2
Chromosome Location:
Chr.5: 135994800 - 136032881 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01201640_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dtx2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256096.1 NP_001243025.1 5 - 6 1413 73
NM_023742.2 NP_076231.1 5 - 6 1413 73
GenBank mRNA AB015423.1 - 5 - 6 1336 73
AK012137.1 - 5 - 6 1409 73
AK173192.1 - 1 - 2 2300 73
AK213536.1 - 1 - 2 88 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001594.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dtx2

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

Notch signaling pathway
protein binding
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500014389.mm.1 TC0500001594.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500014389.mm.1 NM_001256096.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500014389.mm.1 NM_001256096.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500014389.mm.1 NM_023742.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500014389.mm.1 NM_023742.2 Refseq
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 549711 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-z47m5:80/100.66.77.18:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0