This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01202078_g1
See other GFI1 GE Assays ›
Gene Symbol:
Gfi1
Gene Name:
growth factor independent 1
Gene Aliases:
AW495828, Gfi-1, Pal-1, Pal1
Chromosome Location:
Chr.5: 107716655 - 107726031 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01202078_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gfi1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001267621.1 NP_001254550.1 2 - 3 407 135
NM_010278.2 NP_034408.1 1 - 2 619 135
GenBank mRNA AK089065.1 - 2 - 3 408 135
BC130219.1 - 1 - 2 237 135
U58972.1 - 1 - 2 594 135
U78312.1 - 1 - 2 619 135

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC130219.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gfi1

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
mechanosensory behavior
negative regulation of cell fate specification
negative regulation of calcidiol 1-monooxygenase activity
negative regulation of neuron projection development
myeloid cell differentiation
negative regulation of NF-kappaB transcription factor activity
regulation of toll-like receptor signaling pathway
inner ear morphogenesis
auditory receptor cell differentiation
positive regulation of cell fate specification
cell fate commitment
negative regulation of transcription, DNA-templated
regulation of neurogenesis
positive regulation of interleukin-6-mediated signaling pathway
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
nucleic acid binding
DNA binding
protein binding
zinc ion binding
transcription regulatory region DNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0