This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01202296_g1
See other EIF2AK1 GE Assays ›
Gene Symbol:
Eif2ak1
Gene Name:
eukaryotic translation initiation factor 2 alpha kinase 1
Gene Aliases:
HCR, Hri
Chromosome Location:
Chr.5: 143871862 - 143902717 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01202296_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif2ak1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013557.2 NP_038585.2 1538 58
GenBank mRNA AK032508.1 - 109 58
AK156492.1 - 1497 58
AK173160.1 - 1488 58
AK184294.1 - 3 - 4 239 58
AY033898.1 - 1523 58
BC028923.1 - 1510 4164417 58
BC111035.1 - 1489 5357121 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001741.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif2ak1

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

acute inflammatory response
regulation of translation
protein phosphorylation
phagocytosis
negative regulation of cell proliferation
regulation of translational initiation by eIF2 alpha phosphorylation
regulation of eIF2 alpha phosphorylation by heme
phosphorylation
negative regulation of translation
macrophage differentiation
protoporphyrinogen IX metabolic process
regulation of hemoglobin biosynthetic process
negative regulation of hemoglobin biosynthetic process
iron ion homeostasis
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
eukaryotic translation initiation factor 2alpha kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
heme binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500016021.mm.1 TC0500001741.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500016021.mm.1 NM_013557.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500016021.mm.1 NM_013557.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0