This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01202348_m1
See other CHFR GE Assays ›
Gene Symbol:
Chfr
Gene Name:
checkpoint with forkhead and ring finger domains
Gene Aliases:
5730484M20Rik, C230082M18, RNF116
Chromosome Location:
Chr.5: 110135759 - 110171973 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01202348_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Chfr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289577.1 NP_001276506.1 10 - 11 1379 76
NM_001289578.1 NP_001276507.1 9 - 10 1163 76
NM_001289579.1 NP_001276508.1 11 - 12 1452 76
NM_001289580.1 NP_001276509.1 11 - 12 1549 76
NM_172717.4 NP_766305.2 10 - 11 1379 76
GenBank mRNA AK048930.1 - 11 - 12 1530 76
AK077629.1 - 10 - 11 1351 76
AK133154.1 - 10 - 11 1279 76
AK147892.1 - 9 - 10 1138 76
AK148436.1 - 10 - 11 1350 76
AK154038.1 - 10 - 11 1361 76
AK155295.1 - 10 - 11 1351 76
AK155525.1 - 10 - 11 1351 76
AK169182.1 - 11 - 12 1405 76
AK169193.1 - 10 - 11 1334 76
BC049792.1 - 10 - 11 1333 6510480 76

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Chfr

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
mitotic cell cycle
ubiquitin-dependent protein catabolic process
cell cycle
mitotic nuclear division
mitotic cell cycle checkpoint
protein ubiquitination
modification-dependent protein catabolic process
positive regulation of protein ubiquitination
protein destabilization
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
cell division
nucleotide binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
ubiquitin protein ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0