This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01202355_m1
See other CHFR GE Assays ›
Gene Symbol:
Chfr
Gene Name:
checkpoint with forkhead and ring finger domains
Gene Aliases:
5730484M20Rik, C230082M18, RNF116
Chromosome Location:
Chr.5: 110135759 - 110171973 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01202355_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Chfr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289577.1 NP_001276506.1 6 - 7 737 78
NM_001289579.1 NP_001276508.1 7 - 8 810 78
NM_001289580.1 NP_001276509.1 7 - 8 907 78
NM_172717.4 NP_766305.2 6 - 7 737 78
GenBank mRNA AK048930.1 - 7 - 8 887 78
AK052473.1 - 6 - 7 692 78
AK077629.1 - 6 - 7 709 78
AK133154.1 - 7 - 8 792 78
AK148436.1 - 6 - 7 708 78
AK154038.1 - 6 - 7 719 78
AK155295.1 - 6 - 7 709 78
AK155525.1 - 6 - 7 709 78
AK169182.1 - 7 - 8 763 78
AK169193.1 - 6 - 7 692 78
AK196958.1 - 3 - 4 333 78
BC049792.1 - 6 - 7 691 6510480 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001132.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Chfr

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
mitotic cell cycle
ubiquitin-dependent protein catabolic process
cell cycle
mitotic nuclear division
mitotic cell cycle checkpoint
protein ubiquitination
modification-dependent protein catabolic process
positive regulation of protein ubiquitination
protein destabilization
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
cell division
nucleotide binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
ubiquitin protein ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500009654.mm.1 TC0500001132.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500009654.mm.1 NM_001289577.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009654.mm.1 NM_001289579.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009654.mm.1 NM_001289580.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009654.mm.1 NM_172717.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500009655.mm.1 TC0500001132.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500009655.mm.1 NM_001289577.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009655.mm.1 NM_001289578.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009655.mm.1 NM_001289579.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009655.mm.1 NM_001289580.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009655.mm.1 NM_172717.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0