This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01202371_g1
See other P2RX2 GE Assays ›
Gene Symbol:
P2rx2
Gene Name:
purinergic receptor P2X, ligand-gated ion channel, 2
Gene Aliases:
P2X2a, P2x2
Chromosome Location:
Chr.5: 110339812 - 110343446 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01202371_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P2rx2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164833.1 NP_001158305.1 11 - 12 1220 104
GenBank mRNA AB094664.1 - 11 - 12 1140 104

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002988.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Ion Channel 3' Most Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
detection of hypoxic conditions in blood by carotid body chemoreceptor signaling
transport
ion transport
chemical synaptic transmission
neuromuscular synaptic transmission
neuromuscular junction development
sensory perception of sound
response to carbohydrate
response to organic substance
urinary bladder smooth muscle contraction
neuronal action potential
peristalsis
response to ATP
purinergic nucleotide receptor signaling pathway
behavioral response to pain
skeletal muscle fiber development
sensory perception of taste
protein homooligomerization
protein heterooligomerization
cation transmembrane transport
purinergic nucleotide receptor activity
extracellular ATP-gated cation channel activity
ion channel activity
copper ion binding
ATP binding
drug binding
zinc ion binding
ligand-gated ion channel activity
nickel cation binding
phosphatidylinositol binding
identical protein binding
mercury ion binding
cadmium ion binding
cobalt ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500026428.mm.1 TC0500002988.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500026428.mm.1 NM_001164833.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026428.mm.1 NM_001164833.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026428.mm.1 NM_153400.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026428.mm.1 NM_153400.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0