This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01202638_g1
See other GTF2H1 GE Assays ›
Gene Symbol:
Gtf2h1
Gene Name:
general transcription factor II H, polypeptide 1
Gene Aliases:
62kDa, AW743425, AW822074, BTF2 p62, C77871, p62
Chromosome Location:
Chr.7: 46795812 - 46823800 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01202638_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf2h1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291075.1 NP_001278004.1 9 - 10 1142 95
NM_008186.4 NP_032212.3 8 - 9 1175 95
GenBank mRNA AK005065.1 - 8 - 9 1167 95
BC052837.1 - 9 - 10 1154 30049274 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000879.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gtf2h1

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
nucleotide-excision repair
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein phosphorylation
cellular response to DNA damage stimulus
positive regulation of transcription from RNA polymerase II promoter
chromatin binding
protein kinase activity
DNA-dependent ATPase activity
RNA polymerase II carboxy-terminal domain kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700007898.mm.1 NM_008186.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007898.mm.1 NM_008186.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700007898.mm.1 TC0700000879.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700007899.mm.1 NM_008186.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007899.mm.1 NM_008186.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700007899.mm.1 TC0700000879.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline