This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01203485_m1
See other SPTY2D1 GE Assays ›
Gene Symbol:
Spty2d1
Gene Name:
SPT2, Suppressor of Ty, domain containing 1 (S. cerevisiae)
Gene Aliases:
5830435K17Rik, AI852426, P16H6
Chromosome Location:
Chr.7: 46990396 - 47008414 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01203485_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Spty2d1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175318.4 NP_780527.2 2 - 3 455 90
GenBank mRNA AK030869.1 - 2 - 3 290 90
AK147856.1 - 2 - 3 264 90
AK162467.1 - 2 - 3 270 90
BC079859.1 - 2 - 3 250 6834544 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700003049.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Spty2d1

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700027894.mm.1 NM_175318.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700027894.mm.1 NM_175318.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700027894.mm.1 TC0700003049.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0