This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01203623_g1
See other GSTM4 GE Assays ›
Gene Symbol:
Gstm4
Gene Name:
glutathione S-transferase, mu 4
Gene Aliases:
1110004G14Rik, GSTM7-7, Gstb-4, Gstb4
Chromosome Location:
Chr.3: 108040408 - 108045080 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01203623_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gstm4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001160411.1 NP_001153883.1 5 - 6 426 69
NM_026764.3 NP_081040.1 6 - 7 631 69
GenBank mRNA AF464943.1 - 6 - 7 461 69
AF501320.1 - 6 - 7 447 69
AK003418.1 - 5 - 6 427 69
BC030444.1 - 6 - 7 578 5373062 69

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gstm4

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
nitrobenzene metabolic process
xenobiotic catabolic process
glutathione transferase activity
enzyme binding
protein homodimerization activity
glutathione binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300021359.mm.1 NM_026764.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021359.mm.1 NM_026764.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300021359.mm.1 TC0300002728.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300021353.mm.1 NM_001160411.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021353.mm.1 NM_001160411.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021353.mm.1 NM_026764.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021353.mm.1 NM_026764.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300021353.mm.1 TC0300002728.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300021354.mm.1 NM_001160411.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021354.mm.1 NM_001160411.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021354.mm.1 NM_026764.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021354.mm.1 NM_026764.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300021354.mm.1 TC0300002728.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300021350.mm.1 NM_001160411.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021350.mm.1 NM_001160411.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021350.mm.1 NM_026764.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021350.mm.1 NM_026764.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300021350.mm.1 TC0300002728.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300021358.mm.1 NM_026764.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021358.mm.1 NM_026764.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300021358.mm.1 TC0300002728.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300021351.mm.1 NM_001160411.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021351.mm.1 NM_001160411.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021351.mm.1 NM_026764.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021351.mm.1 NM_026764.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300021351.mm.1 TC0300002728.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300021356.mm.1 NM_001160411.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021356.mm.1 NM_001160411.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021356.mm.1 NM_026764.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021356.mm.1 NM_026764.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300021356.mm.1 TC0300002728.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300021357.mm.1 NM_026764.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021357.mm.1 NM_026764.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300021357.mm.1 TC0300002728.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300021352.mm.1 NM_001160411.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021352.mm.1 NM_001160411.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021352.mm.1 NM_026764.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021352.mm.1 NM_026764.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300021352.mm.1 TC0300002728.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300021355.mm.1 NM_001160411.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021355.mm.1 NM_001160411.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021355.mm.1 NM_026764.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021355.mm.1 NM_026764.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300021355.mm.1 TC0300002728.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0