This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01204106_m1
See other HECTD2 GE Assays ›
Gene Symbol:
Hectd2
Gene Name:
HECT domain containing 2
Gene Aliases:
4921524L07, A630025O09Rik, AW212605
Chromosome Location:
Chr.19: 36553969 - 36621135 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01204106_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hectd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172637.3 NP_766225.2 10 - 11 1232 132
GenBank mRNA AK029549.1 - 10 - 11 1092 132

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000520.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hectd2

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination involved in ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900004598.mm.1 NM_001163471.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004598.mm.1 NM_001163471.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004598.mm.1 NM_172637.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004598.mm.1 NM_172637.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900004598.mm.1 TC1900000520.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900004599.mm.1 NM_001163471.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004599.mm.1 NM_001163471.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004599.mm.1 NM_172637.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004599.mm.1 NM_172637.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900004599.mm.1 TC1900000520.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0