This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01204981_m1
See other LIPF GE Assays ›
Gene Symbol:
Lipf
Gene Name:
lipase, gastric
Gene Aliases:
2310051B21Rik, AV082900
Chromosome Location:
Chr.19: 33919441 - 33976813 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01204981_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lipf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026334.3 NP_080610.1 1 - 2 42 87
GenBank mRNA AK009300.1 - 1 - 2 44 87
AK009359.1 - 1 - 2 44 87
AK009428.1 - 1 - 2 46 87
AK009431.1 - 1 - 2 46 87
AK009437.1 - 1 - 2 44 87
AK009459.1 - 1 - 2 44 87
AK009473.1 - 1 - 2 43 87
AK009474.1 - 1 - 2 44 87
AK009479.1 - 1 - 2 41 87
AK009523.1 - 1 - 2 44 87
AK009525.1 - 1 - 2 44 87
AK009537.1 - 1 - 2 44 87
AK009544.1 - 1 - 2 39 87
AK009546.1 - 1 - 2 47 87
AK009560.1 - 1 - 2 44 87
AK009573.1 - 1 - 2 47 87
AK009729.1 - 1 - 2 44 87
AK009773.1 - 1 - 2 44 87
AK009875.1 - 1 - 2 44 87
AK009923.1 - 1 - 2 106 87
AK009990.1 - 1 - 2 44 87
AK010019.1 - 1 - 2 87
AK010026.1 - 1 - 2 44 87
AK010035.1 - 1 - 2 44 87
AK010036.1 - 1 - 2 42 87
AK010058.1 - 1 - 2 43 87
AK010061.1 - 1 - 2 44 87
AK010091.1 - 1 - 2 43 87
AK010093.1 - 1 - 2 44 87
AK010103.1 - 1 - 2 45 87
AK010106.1 - 1 - 2 44 87
AK010110.1 - 1 - 2 47 87
AK010125.1 - 1 - 2 44 87
AK010139.1 - 1 - 2 44 87
AK010148.1 - 1 - 2 44 87
AK010173.1 - 1 - 2 44 87
AK010203.1 - 1 - 2 45 87
AK075913.1 - 1 - 2 45 87
BC061067.1 - 1 - 2 42 30301084 87

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK009300.1, AK009359.1, AK009428.1, AK009431.1, AK009437.1, AK009459.1, AK009473.1, AK009474.1, AK009479.1, AK009523.1, AK009525.1, AK009537.1, AK009544.1, AK009546.1, AK009560.1, AK009573.1, AK009729.1, AK009773.1, AK009875.1, AK009990.1, AK010019.1, AK010026.1, AK010035.1, AK010058.1, AK010061.1, AK010093.1, AK010103.1, AK010106.1, AK010125.1, AK010139.1, AK010148.1, AK010173.1, AK010203.1, BC061067.1, NM_026334.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lipf

Panther Classification:

Molecular Function -

hydrolase lipase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
lipid catabolic process
oxidation-reduction process
triglyceride lipase activity
hydrolase activity
hydrolase activity, acting on ester bonds

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900004349.mm.1 NM_026334.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004349.mm.1 NM_026334.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900004349.mm.1 TC1900000488.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900004350.mm.1 NM_026334.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004350.mm.1 NM_026334.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900004350.mm.1 TC1900000488.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 926942 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hhcgv:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0