This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01204982_g1
See other LIPF GE Assays ›
Gene Symbol:
Lipf
Gene Name:
lipase, gastric
Gene Aliases:
2310051B21Rik, AV082900
Chromosome Location:
Chr.19: 33919441 - 33976813 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01204982_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lipf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026334.3 NP_080610.1 2 - 3 150 131
GenBank mRNA AK009300.1 - 2 - 3 152 131
AK009359.1 - 2 - 3 152 131
AK009413.1 - 2 - 3 151 131
AK009428.1 - 2 - 3 154 131
AK009431.1 - 2 - 3 154 131
AK009437.1 - 2 - 3 152 131
AK009459.1 - 2 - 3 152 131
AK009473.1 - 2 - 3 151 131
AK009474.1 - 2 - 3 152 131
AK009479.1 - 2 - 3 149 131
AK009523.1 - 2 - 3 152 131
AK009525.1 - 2 - 3 152 131
AK009537.1 - 2 - 3 152 131
AK009544.1 - 2 - 3 147 131
AK009546.1 - 2 - 3 155 131
AK009560.1 - 2 - 3 152 131
AK009571.1 - 2 - 3 151 131
AK009573.1 - 2 - 3 155 131
AK009729.1 - 2 - 3 152 131
AK009773.1 - 2 - 3 152 131
AK009875.1 - 2 - 3 152 131
AK009923.1 - 2 - 3 214 131
AK009990.1 - 2 - 3 152 131
AK010019.1 - 2 - 3 153 131
AK010026.1 - 2 - 3 152 131
AK010035.1 - 2 - 3 152 131
AK010036.1 - 2 - 3 150 131
AK010058.1 - 2 - 3 151 131
AK010061.1 - 2 - 3 152 131
AK010091.1 - 2 - 3 151 131
AK010093.1 - 2 - 3 152 131
AK010103.1 - 2 - 3 153 131
AK010106.1 - 2 - 3 152 131
AK010110.1 - 2 - 3 155 131
AK010116.1 - 2 - 3 150 131
AK010123.1 - 2 - 3 153 131
AK010124.1 - 1 - 2 146 131
AK010125.1 - 2 - 3 152 131
AK010139.1 - 2 - 3 152 131
AK010148.1 - 2 - 3 152 131
AK010173.1 - 2 - 3 152 131
AK010203.1 - 2 - 3 153 131
AK010231.1 - 2 - 3 151 131
AK010236.1 - 2 - 3 153 131
AK075913.1 - 2 - 3 153 131
BC061067.1 - 2 - 3 150 30301084 131

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000488.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lipf

Panther Classification:

Molecular Function -

hydrolase lipase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
lipid catabolic process
oxidation-reduction process
triglyceride lipase activity
hydrolase activity
hydrolase activity, acting on ester bonds

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900004352.mm.1 NM_026334.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004352.mm.1 NM_026334.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900004352.mm.1 TC1900000488.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900004351.mm.1 NM_026334.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004351.mm.1 NM_026334.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900004351.mm.1 TC1900000488.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0