This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01204986_m1
See other LIPF GE Assays ›
Gene Symbol:
Lipf
Gene Name:
lipase, gastric
Gene Aliases:
2310051B21Rik, AV082900
Chromosome Location:
Chr.19: 33919441 - 33976813 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01204986_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lipf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026334.3 NP_080610.1 9 - 10 1003 109
GenBank mRNA AK009300.1 - 9 - 10 1005 109
AK009359.1 - 9 - 10 1005 109
AK009413.1 - 9 - 10 1004 109
AK009428.1 - 9 - 10 1007 109
AK009431.1 - 9 - 10 1007 109
AK009437.1 - 9 - 10 1005 109
AK009459.1 - 9 - 10 1005 109
AK009473.1 - 9 - 10 1004 109
AK009474.1 - 9 - 10 1005 109
AK009479.1 - 9 - 10 1002 109
AK009523.1 - 9 - 10 1005 109
AK009525.1 - 9 - 10 1005 109
AK009537.1 - 9 - 10 1005 109
AK009544.1 - 9 - 10 1000 109
AK009546.1 - 9 - 10 1008 109
AK009560.1 - 9 - 10 1005 109
AK009571.1 - 9 - 10 1004 109
AK009573.1 - 9 - 10 1008 109
AK009729.1 - 9 - 10 1005 109
AK009773.1 - 9 - 10 1005 109
AK009875.1 - 9 - 10 1005 109
AK009923.1 - 9 - 10 1066 109
AK010019.1 - 9 - 10 1006 109
AK010026.1 - 9 - 10 1005 109
AK010035.1 - 9 - 10 1005 109
AK010036.1 - 9 - 10 1004 109
AK010058.1 - 9 - 10 1004 109
AK010061.1 - 9 - 10 1005 109
AK010091.1 - 9 - 10 1003 109
AK010093.1 - 9 - 10 1005 109
AK010103.1 - 9 - 10 1006 109
AK010106.1 - 9 - 10 1005 109
AK010110.1 - 9 - 10 1008 109
AK010116.1 - 9 - 10 1003 109
AK010123.1 - 9 - 10 1005 109
AK010124.1 - 8 - 9 999 109
AK010125.1 - 9 - 10 1005 109
AK010139.1 - 9 - 10 1005 109
AK010148.1 - 9 - 10 1005 109
AK010173.1 - 9 - 10 1005 109
AK010203.1 - 9 - 10 1006 109
AK010231.1 - 9 - 10 1004 109
AK010236.1 - 9 - 10 1006 109
AK075910.1 - 2 - 3 119 109
AK075913.1 - 9 - 10 1005 109
BC061067.1 - 9 - 10 1003 30301084 109

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lipf

Panther Classification:

Molecular Function -

hydrolase lipase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
lipid catabolic process
oxidation-reduction process
triglyceride lipase activity
hydrolase activity
hydrolase activity, acting on ester bonds

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P201174 BC061067.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P201174 AK010203.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P201174 AK010231.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P201174 AK010236.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P201174 AK075910.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P201174 BC061067.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P201174 AK010203.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P201174 AK010231.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P201174 AK010236.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P201174 AK075910.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127500 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 385289 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701615 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 771676 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 57756 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 504981 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 129082 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 564689 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 170367 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 101765 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124608 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143290 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P201174 BC061067.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P201174 AK010203.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P201174 AK010231.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P201174 AK010236.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P201174 AK075910.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P201174 BC061067.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P201174 AK010203.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P201174 AK010231.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P201174 AK010236.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P201174 AK075910.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P201174 BC061067.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P201174 AK010203.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P201174 AK010231.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P201174 AK010236.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P201174 AK075910.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P201174 BC061067.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P201174 AK010203.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P201174 AK010231.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P201174 AK010236.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P201174 AK075910.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32077628 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32077634 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32077623 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32077630 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32077629 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32077639 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32077626 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32077620 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32077635 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32077637 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32077618 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32077622 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32077625 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32077636 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32077643 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32077632 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124608 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143290 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09156991 - -
Whole Mouse Genome G4122A A_51_P201174 BC061067.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P201174 AK010203.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P201174 AK010231.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P201174 AK010236.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P201174 AK075910.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P201174 BC061067.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P201174 AK010203.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P201174 AK010231.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P201174 AK010236.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P201174 AK075910.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136529 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103694 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143290 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124608 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14494358 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14494363 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14494357 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077683 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077673 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077650 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077714 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077699 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077651 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077690 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077700 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077722 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077759 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077680 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077706 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077749 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077664 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077657 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077676 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077655 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077730 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077746 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077709 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077754 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077763 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077724 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077625 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077678 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077743 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077728 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077712 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077665 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077732 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077675 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077703 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077693 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077687 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077636 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077639 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077701 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077716 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077753 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077632 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077622 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077729 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077762 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077694 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077719 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077742 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077697 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077757 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077740 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077637 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077721 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077751 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077645 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077684 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077620 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077744 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077761 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077659 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077692 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077629 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077667 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077738 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077663 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077660 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077731 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077669 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077727 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077653 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077670 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32077767 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077718 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077630 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077636 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077658 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077729 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077659 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077721 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077655 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077728 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077628 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077648 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077722 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077670 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077671 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077637 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077650 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077639 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077625 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077675 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14342861 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14336716 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077676 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077618 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077646 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14399470 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077651 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077719 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077632 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14336711 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077643 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14399467 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077667 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077665 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14336697 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077653 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077673 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077716 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077731 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077629 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077664 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14439883 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077622 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077724 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14459729 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077663 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077620 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077647 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077727 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077640 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32077645 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10462544 - -
Affymetrix Mouse Gene 1.0 ST Array 10462546 - -
Affymetrix Mouse Gene 1.0 ST Array 10462550 - -
Affymetrix Mouse Gene 1.0 ST Array 10462551 - -
Affymetrix Mouse Gene 1.0 ST Array 10462549 - -
Affymetrix Mouse Gene 1.0 ST Array 10462548 - -
Affymetrix Mouse Gene 1.0 ST Array 10462547 - -
Affymetrix Mouse Gene 1.0 ST Array 10462552 - -
Affymetrix Mouse Gene 1.0 ST Array 10462545 - -
Affymetrix Mouse Gene 1.0 ST Array 10462553 - -
Affymetrix HT MG-430 PM Array Plate 1416040_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10462542 - -
Mouse Genome 430 2.0 Array 1416040_PM_at - -
Mouse Genome 430 2.0 Array 1416040_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 67717_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 67717_PM_at - -
Mouse Expression Set 430 1416040_PM_at - -
Mouse Expression Set 430 1416040_at - -
Mouse Genome 430A 2.0 Array 1416040_at - -
Mouse Genome 430A 2.0 Array 1416040_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1416040_at - -
Affymetrix HT Mouse Genome 430A Array 1416040_PM_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 5050368 - -
Illumina mouseRef-8 v1.0 expression beadchip 0005050368 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2681664 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2681664 - -
mouse-6 v1.1 (Illumina) scl53201.9.1_2-S - -
Illumina Sentrix mouse-6.v1 bead array 5050368 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl53201.9.1_2-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 5050368 - -
MouseRef-8 v2 Expression BeadChip ILMN_2863532 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2863532 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gsg6q:80/100.66.76.182:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0