This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01205042_m1
See other TRPS1 GE Assays ›
Gene Symbol:
Trps1
Gene Name:
trichorhinophalangeal syndrome I (human)
Gene Aliases:
AI115454, AI447310, D15Ertd586e
Chromosome Location:
Chr.15: 50654757 - 50890747 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01205042_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trps1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032000.2 NP_114389.2 2 - 3 1368 76
GenBank mRNA AF346836.1 - 3 - 4 1293 76
AK142355.1 - 2 - 3 1038 76
BC037058.1 - 2 - 3 1046 4951294 76
BC049857.1 - 4 - 5 934 4460789 76
BC099962.1 - 2 - 3 1333 30091008 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001491.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trps1

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
chondrocyte differentiation
transcription, DNA-templated
regulation of transcription, DNA-templated
transmembrane receptor protein serine/threonine kinase signaling pathway
regulation of histone deacetylation
regulation of chondrocyte differentiation
negative regulation of transcription, DNA-templated
positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding
sequence-specific DNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500011359.mm.1 NM_032000.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500011359.mm.1 NM_032000.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500011359.mm.1 TC1500001491.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500011375.mm.1 NM_032000.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500011375.mm.1 NM_032000.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500011375.mm.1 TC1500001491.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500011378.mm.1 NM_032000.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500011378.mm.1 NM_032000.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500011378.mm.1 TC1500001491.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500011361.mm.1 NM_032000.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500011361.mm.1 NM_032000.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500011361.mm.1 TC1500001491.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500011379.mm.1 NM_032000.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500011379.mm.1 NM_032000.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500011379.mm.1 TC1500001491.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500011360.mm.1 NM_032000.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500011360.mm.1 NM_032000.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500011360.mm.1 TC1500001491.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500011380.mm.1 NM_032000.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500011380.mm.1 NM_032000.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500011380.mm.1 TC1500001491.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0