This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01205388_m1
See other PITPNB GE Assays ›
Gene Symbol:
Pitpnb
Gene Name:
phosphatidylinositol transfer protein, beta
Gene Aliases:
PI-TP-beta
Chromosome Location:
Chr.5: 111330697 - 111388361 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01205388_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pitpnb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301643.1 NP_001288572.1 3 - 4 312 70
NM_001301644.1 NP_001288573.1 3 - 4 281 70
NM_001301666.1 NP_001288595.1 3 - 4 312 70
NM_019640.5 NP_062614.1 3 - 4 312 70
GenBank mRNA AK028235.1 - 3 - 4 247 70
BC034676.1 - 3 - 4 202 4481685 70
BQ178694.1 - 3 - 4 201 70
BY205337.1 - 3 - 4 279 70
CK619028.1 - 3 - 4 195 70
U46934.1 - 3 - 4 214 70

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001301644.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pitpnb

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
transport
phospholipid transport
phospholipid binding
lipid binding
phosphatidylinositol transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500009962.mm.1 NM_019640.5 Refseq
Affymetrix Mouse Clariom S Array PSR0500009962.mm.1 TC0500001162.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500009964.mm.1 NM_019640.5 Refseq
Affymetrix Mouse Clariom S Array PSR0500009964.mm.1 TC0500001162.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline