This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01205449_g1
See other FCGRT GE Assays ›
Gene Symbol:
Fcgrt
Gene Name:
Fc receptor, IgG, alpha chain transporter
Gene Aliases:
FcRn
Chromosome Location:
Chr.7: 45092992 - 45103846 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01205449_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fcgrt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010189.3 NP_034319.2 1 - 2 179 153
GenBank mRNA AK008167.1 - 1 - 2 153 153
AK008360.1 - 1 - 2 153 153
AK146933.1 - 1 - 2 141 154
AK152879.1 - 1 - 2 115 153
BC003786.1 - 1 - 2 102 3485649 153
BY063885.1 - 1 - 2 210 153
D37874.1 - 1 - 2 112 153

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK152879.1, BC003786.1, D37874.1, NM_010189.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fcgrt

Panther Classification:

Molecular Function -

major histocompatibility complex protein

Gene Ontology Categories:

Function(s) Process(es)

immune response
antigen processing and presentation
Fc-gamma receptor signaling pathway
antigen binding
IgG receptor activity
IgG binding
beta-2-microglobulin binding

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 390657 - -
ABI Mouse Genome Survey Microarray 682726 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline