This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01205450_g1
See other FCGRT GE Assays ›
Gene Symbol:
Fcgrt
Gene Name:
Fc receptor, IgG, alpha chain transporter
Gene Aliases:
FcRn
Chromosome Location:
Chr.7: 45092992 - 45103846 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01205450_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fcgrt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010189.3 NP_034319.2 2 - 3 456 71
GenBank mRNA AK146933.1 - 2 - 3 418 71
AK152879.1 - 2 - 3 392 71
BC003786.1 - 2 - 3 379 3485649 71
D37874.1 - 2 - 3 389 71
L17022.1 - 1 - 2 95 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002989.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fcgrt

Panther Classification:

Molecular Function -

major histocompatibility complex protein

Gene Ontology Categories:

Function(s) Process(es)

immune response
antigen processing and presentation
Fc-gamma receptor signaling pathway
antigen binding
IgG receptor activity
IgG binding
beta-2-microglobulin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0