Assay Details
| Gene Symbol: Dhdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027903.3 | NP_082179.1 | 2 - 3 | 238 | 85 | |
| GenBank mRNA | AK005050.2 | - | 2 - 3 | 239 | 85 | |
| AK154452.1 | - | 2 - 3 | 220 | 85 | ||
| BC031710.1 | - | 2 - 3 | 216 | 85 | ||
| BC116414.2 | - | 2 - 3 | 222 | 40103176 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhdh
Genomic Map