This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01205879_g1
See other SIGLECG GE Assays ›
Gene Symbol:
Siglecg
Gene Name:
sialic acid binding Ig-like lectin G
Gene Aliases:
9830164H23, A630096C01Rik, Siglec-G, Siglec10, mSiglec-G
Chromosome Location:
Chr.7: 43408193 - 43418350 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01205879_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Siglecg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172900.3 NP_766488.2 1 - 2 92 63
GenBank mRNA AK036698.1 - 1 - 2 106 63
AK042488.1 - 1 - 2 93 63
AK156766.1 - 1 - 2 134 63

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK036698.1, AK042488.1, AK156766.1, NM_172900.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Siglecg

Panther Classification:

Molecular Function -

immunoglobulin superfamily cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700006098.mm.1 NM_172900.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700006098.mm.1 NM_172900.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700006098.mm.1 TC0700000757.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700006100.mm.1 NM_172900.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700006100.mm.1 NM_172900.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700006100.mm.1 TC0700000757.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline