This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01205883_g1
See other SIGLECG GE Assays ›
Gene Symbol:
Siglecg
Gene Name:
sialic acid binding Ig-like lectin G
Gene Aliases:
9830164H23, A630096C01Rik, Siglec-G, Siglec10, mSiglec-G
Chromosome Location:
Chr.7: 43408193 - 43418350 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01205883_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Siglecg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172900.3 NP_766488.2 10 - 11 1829 76
GenBank mRNA AK036698.1 - 7 - 8 3285 76
AK042488.1 - 10 - 11 1830 76
AK156766.1 - 6 - 7 3238 76
AY210400.1 - 9 - 10 1684 76
EF419770.1 - 9 - 10 1692 76

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Siglecg

Panther Classification:

Molecular Function -

immunoglobulin superfamily cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline